Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse GRIN1/NMDAR1 Alexa Fluor™ 647-conjugated Antibody, R&D Systems™

Rat Monoclonal Antibody
Supplier: R&D Systems IC10655R100UG
This item is not returnable.
View return policy
Description
NMDA R, NR1 Subunit Monoclonal antibody specifically detects NMDA R, NR1 Subunit in Mouse samples. It is validated for Flow CytometrySpecifications
NMDA R, NR1 Subunit | |
Monoclonal | |
Alexa Fluor 647 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
Rat | |
Protein A or G purified from cell culture supernatant | |
RUO | |
2902 | |
Mouse | |
Purified |
Flow Cytometry | |
1031915 | |
Intracellular Staining by Flow Cytometry 0.25-1 ug/106 cells | |
GluN1, MRD8, NDHMSD, NMDA R, NR1 Subunit, NMDA1, NMDAR1, NMD-R1, NR1 | |
Mouse myeloma cell line NS0-derived recombinant mouse GRIN1/NMDAR1, Met1-Gln559 | |
100 μg | |
Primary | |
Reconstitution default not known | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction