Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse GRIN1/NMDAR1 Antibody, R&D Systems™

Rat Monoclonal Antibody
Supplier: R&D Systems MAB10655100
This item is not returnable.
View return policy
Description
NMDA R, NR1 Subunit Monoclonal antibody specifically detects NMDA R, NR1 Subunit in Mouse samples. It is validated for Flow Cytometry, ImmunocytochemistrySpecifications
NMDA R, NR1 Subunit | |
Monoclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
Rat | |
Protein A or G purified from cell culture supernatant | |
RUO | |
2902 | |
Mouse | |
Purified |
Flow Cytometry, Immunocytochemistry | |
1031915 | |
Intracellular Staining by Flow Cytometry 0.25 ug/106 cells, Immunocytochemistry 8-25 ug/mL | |
GluN1, MRD8, NDHMSD, NMDA R, NR1 Subunit, NMDA1, NMDAR1, NMD-R1, NR1 | |
Mouse myeloma cell line NS0-derived recombinant mouse GRIN1/NMDAR1, Met1-Gln559 | |
100 μg | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction