Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Mouse GSTA1 ELISA Kit (Colorimetric)
SDP

Catalog No. NB170568 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB170568 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB170568 Supplier Novus Biologicals™ Supplier No. NBP318721
Add to Cart
Add to Cart
This item is not returnable. View return policy

Colorimetric ELISA Kit

Specifications

Assay Range 0.25 - 16 ng/mL (example only; lot dependent)
Assay Sensitivity 0.19 ng/mL (example only; lot dependent)
Product Type ELISA Kit (Colorimetric)
Sample Type Cell Culture, Plasma, Serum, Tissue Extracts, Urine
For Use With (Application) ELISA
Gene ID (Entrez) 2938
Gene Symbol GSTA1
Quantity 1 Kit
Research Discipline Cellular Signaling, Epitope Tags, metabolism
Storage Requirements Storage is content dependent.
Synonym EC 2.5.1.18, glutathione S-alkyltransferase A1, glutathione S-aryltransferase A1, glutathione S-transferase 2, glutathione S-transferase A1, glutathione S-transferase alpha 1, glutathione S-transferase Ha subunit 1, GST class-alpha member 1, GST HA subunit 1, GST, class alpha, 1, GST2, GSTA1-1, GST-epsilon, GTH1, MGC131939, S-(hydroxyalkyl)glutathione lyase A1
Target Mouse
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.