Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse/Hamster GDF-9 Antibody, R&D Systems™
Goat Polyclonal Antibody has been used in 2 publications
Supplier: R&D Systems AF739SP
This item is not returnable.
View return policy
Description
GDF-9 Polyclonal specifically detects GDF-9 in Mouse, Hamster samples. It is validated for Western Blot, Immunohistochemistry.Specifications
GDF-9 | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
GDF9 | |
E. coli-derived recombinant mouse GDF-9 | |
25 μg | |
Primary | |
Detects mouse GDF-9 in direct ELISAs and both mouse and hamster GDF-9 in Western blots. In direct ELISAs, less than 1% cross-reactivity with recombinant mouse (rm) GDF-1, rmGDF-5, rmGDF-6, rmGDF-8, and rmGDF-15 is observed. | |
Mouse, Hamster | |
IgG |
Western Blot, Immunohistochemistry | |
LYOPH | |
Western Blot 1 ug/mL, Immunohistochemistry 5-15 ug/mL | |
GDF9, GDF-9, growth differentiation factor 9, growth/differentiation factor 9 | |
Goat | |
Affinity Purified | |
RUO | |
2661 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction