Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse HB-EGF Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF8239
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF8239 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF8239 Supplier R&D Systems Supplier No. AF8239
Add to Cart
Add to Cart
This item is not returnable. View return policy

Sheep Polyclonal Antibody has been used in 4 publications

HB-EGF Polyclonal specifically detects HB-EGF in Mouse samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen HB-EGF
Applications Western Blot, Immunohistochemistry
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 2 ug/mL, Immunohistochemistry 5-15 ug/mL
Gene Accession No. Q06186
Gene Alias diphtheria toxin receptor (heparin-binding epidermal growth factor-like growthfactor), Dtr, DTRHEGFLdiphtheria toxin receptor (heparin-binding EGF-like growth factor), Dts, DTSF, HBEGF, Hegfl, heparin-binding EGF-like growth factor, heparin-binding epidermal growth factor, proheparin-binding EGF-like growth factor
Gene Symbols HBEGF
Host Species Sheep
Immunogen E. coli-derived recombinant mouse HB-EGF Asp63-Leu148 Accession # Q06186
Purification Method Affinity Purified
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 1839
Test Specificity Detects mouse HB-EGF in direct ELISAs and Western Blots. In direct ELISAs, less than 30% cross-reactivity with recombinant human HB-EGF is observed and less then 2% cross-reactivity with recombinant mouse Amphiregulin is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.