Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse Hematopoietic Lineage Marker Alexa Fluor 405 Antibody Cocktail, R&D Systems™

Rat Monoclonal Antibody cocktail
Supplier: R&D Systems FLC001V
Includes: 500μL of an AlexaFluor 405-conjugated antibody cocktail of the following mouse lineage markers:
- Anti-mouse CD3 (Clone 17A2)Anti-mouse B220 (Clone RA3-6B2)Anti-mouse CD11b (Clone M1/70)Anti-mouse GR-1 (Clone RB6-8C5)Anti-mouse Ter-119 (Clone TER119)Anti-mouse CD5 (Clone 53-7.3)
Description
Hematopoietic Lineage Marker Monoclonal specifically detects Hematopoietic Lineage Marker in Mouse samples. It is validated for Flow Cytometry.Specifications
Hematopoietic Lineage Marker | |
Monoclonal | |
Alexa Fluor 405 | |
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
Unpurified | |
RUO | |
Mouse | |
IgG2b |
Flow Cytometry | |
17A2, RA3-6B2, M1/70, RB6-8C5, TER119, 53-7.3 | |
Flow Cytometry | |
Rat | |
25 Tests | |
Primary | |
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction