Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse Hematopoietic Lineage Marker Allophycocyanin Antibody Cocktail, R&D Systems™
GREENER_CHOICE

Catalog No. FLC001A
Change view
Click to view available options
Quantity:
25 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FLC001A 25 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FLC001A Supplier R&D Systems Supplier No. FLC001A
Add to Cart
Add to Cart
This item is not returnable. View return policy

Includes

500μL of an FITC-conjugated antibody cocktail of the following mouse lineage markers:

  • Anti-mouse CD3 (Clone 17A2)Anti-mouse B220 (Clone RA3-6B2)Anti-mouse CD11b (Clone M1/70)Anti-mouse GR-1 (Clone RB6-8C5)Anti-mouse Ter-119 (Clone TER119)Anti-mouse CD5 (Clone 53-7.3)

Rat Monoclonal Antibody cocktail

Hematopoietic Lineage Marker Monoclonal specifically detects Hematopoietic Lineage Marker in Mouse samples. It is validated for Flow Cytometry.

Specifications

Antigen Hematopoietic Lineage Marker
Applications Flow Cytometry
Classification Monoclonal
Clone 17A2, RA3-6B2, M1/70, RB6-8C5, TER119, 53-7.3
Conjugate APC
Dilution Flow Cytometry
Formulation Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Host Species Rat
Purification Method Unpurified
Quantity 25 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.