Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse Hemopexin Antibody, R&D Systems™

Rat Monoclonal Antibody
Supplier: R&D Systems MAB7007SP
This item is not returnable.
View return policy
Description
Hemopexin Monoclonal specifically detects Hemopexin in Mouse samples. It is validated for Western Blot.Specifications
Hemopexin | |
Monoclonal | |
LYOPH | |
Western Blot 2 ug/mL | |
Q91X72 | |
HPX | |
Mouse myeloma cell line NS0-derived recombinant mouse Hemopexin Met1-Gln460 Accession # Q91X72 | |
25 μg | |
Primary | |
Detects mouse Hemopexin in direct ELISAs and Western blots. In direct ELISAs, less than 5% cross-reactivity with recombinant human Hemopexin is observed. | |
Mouse | |
Purified |
Western Blot | |
781941 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
beta-1B-glycoprotein, FLJ56652, hemopexin, HPX, HX | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
3263 | |
Sterile PBS to a final concentration of 0.5 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction