Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse Hepcidin Antibody, R&D Systems™

Rat Monoclonal Antibody
Supplier: R&D Systems MAB9505100
This item is not returnable.
View return policy
Description
Hepcidin Antimicrobial Peptide Monoclonal specifically detects Hepcidin Antimicrobial Peptide in Mouse samples. It is validated for ELISA.Specifications
HepcidinAntimicrobialPeptide | |
Monoclonal | |
Unconjugated | |
Lyophilized from a 0.2μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2μm filtered solution in PBS. | |
HAMP, HEPC, hepcidin antimicrobial peptide, HEPCPutative liver tumor regressor, HFE2B, HFE2Bhepcidin, LEAP1, LEAP-1, LEAP1PLTR, Liver-expressed antimicrobial peptide 1, PLTR | |
Synthetic peptide containing mouse Hepcidin Accession # Q9EQ21 | |
100 μg | |
57817 | |
Reconstitute at 0.5mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2a |
ELISA | |
968001 | |
ELISA | |
Q9EQ21 | |
Rat | |
Protein A or G purified from cell culture supernatant | |
Primary | |
Detects mouse Hepcidin in direct ELISA. | |
Mouse | |
Supernatant |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction