Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse HGFR/c-MET Biotinylated Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 2 publications
Supplier: R&D Systems BAF527
This item is not returnable.
View return policy
Description
HGFR/c-MET Polyclonal specifically detects HGFR/c-MET in Mouse samples. It is validated for Western Blot, Immunohistochemistry, ELISA Detection (Matched Antibody Pair).Specifications
HGFR/c-MET | |
Polyclonal | |
Biotin | |
P16056 | |
MET | |
S. frugiperda insect ovarian cell line Sf 21-derived recombinant mouse HGF R/c-MET Glu25-Asn929 Accession # P16056 | |
50 μg | |
Primary | |
Detects mouse HGF R/c-MET in ELISAs and Western blots. In sandwich immunoassays, less than 0.2% cross-reactivity with recombinant human HGF R, recombinant mouse (rm) HGF A, and rmMSP R is observed. | |
Mouse | |
IgG |
Western Blot, Immunohistochemistry, ELISA | |
LYOPH | |
Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL, ELISA Detection (Matched Antibody Pair) 0.1-0.4 ug/mL | |
AUTS9, c-Met, cMET, EC 2.7.10, EC 2.7.10.1, hepatocyte growth factor receptor, HGF R, HGF receptor, HGF/SF receptor, HGFR, MET, Met (c-Met), met proto-oncogene (hepatocyte growth factor receptor), met proto-oncogene tyrosine kinase, oncogene MET, Proto-oncogene c-Met, RCCP2, Scatter factor receptor, SF receptor, Tyrosine-protein kinase Met | |
Goat | |
Affinity Purified | |
RUO | |
4233 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction