Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse HRP-1 Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems AF1868
This item is not returnable.
View return policy
Description
HRP-1/HDGFL1 Polyclonal specifically detects HRP-1/HDGFL1 in Mouse samples. It is validated for Western Blot.Specifications
HRP-1/HDGFL1 | |
Polyclonal | |
Unconjugated | |
NP_032258 | |
HDGFL1 | |
E. coli-derived recombinant human HRP-1 Ser2-Leu283 Accession # NP_032258 | |
100 μg | |
Primary | |
Detects mouse HRP-1 in direct ELISAs and Western blots. | |
Mouse | |
IgG |
Western Blot | |
LYOPH | |
Western Blot 0.1 ug/mL | |
dJ309H15.1, HDGF (hepatoma-derived growth factor) like, hepatoma derived growth factor-like 1, hepatoma-derived growth factor-like protein 1, HRP1, PWWP domain containing 1, PWWP domain-containing protein 1, PWWP1 | |
Goat | |
Affinity Purified | |
RUO | |
154150 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction