Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse HSF1 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB7130
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB7130 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB7130 Supplier R&D Systems Supplier No. MAB7130
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

HSF1 Monoclonal specifically detects HSF1 in Mouse samples. It is validated for Immunohistochemistry.

Specifications

Antigen HSF1
Applications Immunohistochemistry
Classification Monoclonal
Clone 744509
Concentration LYOPH
Conjugate Unconjugated
Dilution Immunohistochemistry 8-25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P38532
Gene Alias heat shock factor protein 1, heat shock transcription factor 1HSTF1HSF 1, HSTF 1, HSTF1
Gene Symbols HSF1
Host Species Rat
Immunogen E. coli-derived recombinant mouse HSF1 Thr124-Tyr247 Accession # P38532
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3297
Test Specificity Detects mouse HSF1 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human HSF2 or recombinant mouse HSF4 is observed.
Reconstitution Sterile PBS to a final concentration of 0.5 mg/mL.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.