Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse HSF1 Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems AF7130SP
This item is not returnable.
View return policy
Description
HSF1 Polyclonal specifically detects HSF1 in Mouse samples. It is validated for Immunohistochemistry.Specifications
HSF1 | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
heat shock factor protein 1, heat shock transcription factor 1HSTF1HSF 1, HSTF 1, HSTF1 | |
Goat | |
Affinity Purified | |
RUO | |
3297 | |
Sterile PBS to a final concentration of 0.2 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Immunohistochemistry | |
LYOPH | |
Immunohistochemistry 5-15 ug/mL | |
P38532 | |
HSF1 | |
E. coli-derived recombinant mouse HSF1 Thr124-Tyr247 Accession # P38532 | |
25 μg | |
Primary | |
Detects mouse HSF1 in direct ELISAs. In direct ELISAs, approximately 6% cross-reactivity with recombinant mouse HSF4 is observed and less than 1% cross-reactivity with recombinant human HSF2 is observed. | |
Mouse | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction