Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse HSF4 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF4088SP
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF4088SP 25 μg
AF4088 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. AF4088SP Supplier R&D Systems Supplier No. AF4088SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

HSF4 Polyclonal specifically detects HSF4 in Mouse samples. It is validated for Western Blot.

Specifications

Antigen HSF4
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.1 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q9R0L1
Gene Alias ATHSF4, AT-HSFB1, cataract, Marner, CTM, heat shock factor protein 4, heat shock transcription factor 4CTM, hHSF4, HSF 4, HSFB1, HSTF 4
Gene Symbols HSF4
Host Species Goat
Immunogen E. coli-derived recombinant mouse HSF4 Gly343-Pro492 Accession # Q9R0L1
Purification Method Affinity Purified
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3299
Test Specificity Detects mouse HSF4 in direct ELISAs and Western blots. In Western blots, approximately 25% cross-reactivity with recombinant human (rh) HSF4 is observed and less than 5% cross-reactivity with recombinant mouse HSF1 and rhHSF2 is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.