Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse HVEM/TNFRSF14 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF2516SP
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF2516SP 25 μg
AF2516 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. AF2516SP Supplier R&D Systems Supplier No. AF2516SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

HVEM/TNFRSF14 Polyclonal specifically detects HVEM/TNFRSF14 in Mouse samples. It is validated for Western Blot.

Specifications

Antigen HVEM/TNFRSF14
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.1 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. NP_849262
Gene Alias ATAR, CD270, CD270 antigen, CD40-like protein, Herpes virus entry mediator A, Herpesvirus entry mediator A, HveA, HVEMTR2HVEAATAR, LIGHTR, TNFRSF14, tumor necrosis factor receptor superfamily member 14, tumor necrosis factor receptor superfamily, member 14 (herpesvirus entrymediator), Tumor necrosis factor receptor-like 2, tumor necrosis factor receptor-like gene2
Gene Symbols TNFRSF14
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse HVEM/TNFRSF14 Gln39-Val207 Accession # NP_849262
Purification Method Affinity Purified
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8764
Test Specificity Detects mouse HVEM/TNFRSF14 in direct ELISAs and Western blots. In direct ELISAs, approximately 100% cross-reactivity with recombinant rat HVEM/TNFRSF14 is observed, and less than 5% cross-reactivity with recombinant human (rh) HVEM is observed and less than 2% cross-reactivity with recombinant mouse (rm) DR3, rmFas, rmGITR, rmTWEAK R, rm4-1BB, rmBAFF R, rmCD27, rmCD30, rmCD40, rmEDAR, rmNGF R, rmOX40, rmRANK, rmTNF RI, rmTNF RII, rhDR6, rhTRAIL R3, and rhTRAIL R4 is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.