Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse ICAM-1/CD54 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF796SP
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF796SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF796SP Supplier R&D Systems Supplier No. AF796SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody has been used in 43 publications

ICAM-1/CD54 Polyclonal specifically detects ICAM-1/CD54 in Mouse samples. It is validated for Western Blot, Immunohistochemistry, Adhesion Blockade.

Specifications

Antigen ICAM-1/CD54
Applications Western Blot, Immunohistochemistry, Adhesion Blockade
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL, Adhesion Blockade
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q3U8M7
Gene Alias BB2, CD54, CD54 antigen, cell surface glycoprotein P3.58, human rhinovirus receptor, ICAM1, ICAM-1, intercellular adhesion molecule 1, intercellular adhesion molecule 1 (CD54), human rhinovirus receptor, Major group rhinovirus receptor, P3.58
Gene Symbols ICAM1
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse ICAM-1/CD54 Gln28-Asn485 Accession # Q3U8M7
Purification Method Affinity Purified
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3383
Test Specificity Detects mouse ICAM-1 in direct ELISAs and Western blots. In direct ELISAs, approximately 20% cross-reactivity with recombinant rat ICAM-1 is observed, approximately 5% cross-reactivity with recombinant human (rh) ICAM-1 and rhICAM-3 is observed, and less than 1% cross-reactivity with recombinant mouse ICAM-2 is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.