Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse ICAM-1/CD54 Biotinylated Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems BAF796
This item is not returnable.
View return policy
Description
ICAM-1/CD54 Polyclonal specifically detects ICAM-1/CD54 in Mouse samples. It is validated for Western Blot, Immunohistochemistry, ELISA Detection (Matched Antibody Pair).Specifications
ICAM-1/CD54 | |
Polyclonal | |
Biotin | |
Q3U8M7 | |
ICAM1 | |
Mouse myeloma cell line NS0-derived recombinant mouse ICAM-1/CD54 Gln28-Asn485 Accession # Q3U8M7 | |
50 μg | |
Primary | |
Detects mouse ICAM-1/CD54 in ELISAs and Western blots. In sandwich ELISAs, less than 0.3% cross-reactivity with recombinant human ICAM-1, recombinant rat ICAM-1, and recombinant mouse ICAM-2 is observed. | |
Mouse | |
IgG |
Western Blot, Immunohistochemistry, ELISA | |
LYOPH | |
Western Blot 0.1 ug/mL, Immunohistochemistry 2-10 ug/mL, ELISA Detection (Matched Antibody Pair) 0.1-0.4 ug/mL | |
BB2, CD54, CD54 antigen, cell surface glycoprotein P3.58, human rhinovirus receptor, ICAM1, ICAM-1, intercellular adhesion molecule 1, intercellular adhesion molecule 1 (CD54), human rhinovirus receptor, Major group rhinovirus receptor, P3.58 | |
Goat | |
Affinity Purified | |
RUO | |
3383 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction