Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse ICAM-1/CD54 PE-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB796P
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB796P 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB796P Supplier R&D Systems Supplier No. FAB796P
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

ICAM-1/CD54 Monoclonal specifically detects ICAM-1/CD54 in Mouse samples. It is validated for Flow Cytometry.

Specifications

Antigen ICAM-1/CD54
Applications Flow Cytometry
Classification Monoclonal
Clone 166623
Conjugate R-PE
Dilution Flow Cytometry 10 uL/10^6 cells
Formulation Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Gene Accession No. P13597
Gene Alias BB2, CD54, CD54 antigen, cell surface glycoprotein P3.58, human rhinovirus receptor, ICAM1, ICAM-1, intercellular adhesion molecule 1, intercellular adhesion molecule 1 (CD54), human rhinovirus receptor, Major group rhinovirus receptor, P3.58
Gene Symbols ICAM1
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse ICAM-1 Gln28-Asn485 Accession # P13597
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3383
Test Specificity Detects mouse ICAM-1 in ELISAs and Western blots. In Western blots, this antibody shows weak cross-reactivity with recombinant human (rh) ICAM-1 and recombinant mouse (rm) ICAM-2 and no cross-reactivity with rhCD31, rmDCC, rhICAM-2, rhICAM-3, recombinant rat (rr) ICAM-1, rmMAdCAM-1, or rmVCAM-1.
Target Species Mouse
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.