Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse ICAM-5 Antibody, R&D Systems™

Rat Monoclonal Antibody
$200.50 - $643.50
Specifications
| Antigen | ICAM-5 |
|---|---|
| Clone | 191321 |
| Concentration | LYOPH |
| Dilution | Western Blot 1 ug/mL |
| Applications | Western Blot |
Description
ICAM-5 Monoclonal specifically detects ICAM-5 in Mouse samples. It is validated for Western Blot.Specifications
| ICAM-5 | |
| LYOPH | |
| Western Blot | |
| Unconjugated | |
| Rat | |
| Mouse | |
| Q60625 | |
| 7087 | |
| Mouse myeloma cell line NS0-derived recombinant mouse ICAM-5 Leu31-Arg828 (Pro47Arg) Accession # Q60625 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects mouse ICAM-5 in direct ELISAs and Western blots. In direct ELISAs and Western blots, this antibody does not cross-react with recombinant mouse (rm) DCC, rmICAM-1, rhICAM-1, rmICAM-2, rhICAM-3, rmMadCAM, rhCD31, or rmVCAM-1. |
| 191321 | |
| Western Blot 1 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| ICAM5, ICAM-5, intercellular adhesion molecule 5, intercellular adhesion molecule 5, telencephalin, Telencephalin, TLCNTelencephalin, TLNtelencephalin | |
| ICAM5 | |
| IgG2a | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title