Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse ICOS Alexa Fluor™ 647-conjugated Antibody, R&D Systems™

Rabbit Monoclonal Antibody
Supplier: R&D Systems FAB1682R100UG
This item is not returnable.
View return policy
Description
ICOS Monoclonal specifically detects ICOS in Mouse samples. It is validated for Flow Cytometry.Specifications
ICOS | |
Monoclonal | |
Alexa Fluor 647 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
Activation-inducible lymphocyte immunomediatory molecule, AILIM, AILIMCD278, CD278, CD278 antigen, CRP-1, CVID1, inducible costimulator, inducible T-cell costimulator, inducible T-cell co-stimulator, MGC39850 | |
Chinese Hamster Ovary cell line, CHO-derived mouse ICOS, Met1-Leu142, Accession # Q9WVS0 | |
100 μg | |
29851 | |
Mouse | |
Supernatant |
Flow Cytometry | |
2317C | |
Flow Cytometry 0.25-1 μg/106 cells | |
Q9WVS0 | |
Rabbit | |
Protein A or G purified from hybridoma culture supernatant | |
Primary | |
Detects mouse ICOS in direct ELISAs. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction