Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse ICOS Antibody, R&D Systems™

Rat Monoclonal Antibody
Supplier: R&D Systems MAB168SP
This item is not returnable.
View return policy
Description
ICOS Monoclonal specifically detects ICOS in Mouse samples. It is validated for Flow Cytometry, CyTOF-ready.Specifications
ICOS | |
Monoclonal | |
LYOPH | |
Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready | |
Q9WVS0 | |
ICOS | |
NS0 mouse myeloma cell line transfected with mouse ICOS Accession # Q9WVS0 | |
25 μg | |
Primary | |
Detects mouse ICOS in direct ELISAs. | |
Mouse | |
Purified |
Flow Cytometry, CyTOF | |
670306 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
Activation-inducible lymphocyte immunomediatory molecule, AILIM, AILIMCD278, CD278, CD278 antigen, CRP-1, CVID1, inducible costimulator, inducible T-cell costimulator, inducible T-cell co-stimulator, MGC39850 | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
29851 | |
Sterile PBS to a final concentration of 0.5 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction