Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse IFITM3/Fragilis Biotinylated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. BAF3377
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BAF3377 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. BAF3377 Supplier R&D Systems Supplier No. BAF3377
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

IFITM3/Fragilis Polyclonal specifically detects IFITM3/Fragilis in Mouse samples. It is validated for Western Blot.

Specifications

Antigen IFITM3/Fragilis
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Biotin
Dilution Western Blot 0.1 ug/mL
Gene Accession No. NP_079654
Gene Alias 1-8U, Fragilis, interferon induced transmembrane protein 3 (1-8U), interferon-induced transmembrane protein 3, Interferon-inducible protein 1-8U, IP15, mil-1
Gene Symbols IFITM3
Host Species Goat
Immunogen E. coli-derived recombinant mouse IFITM3/Fragilis Asn2-His57 Accession # NP_079654
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10410
Test Specificity Detects mouse IFITM3/Fragilis in Western blots. In Western blots, approximately 50% cross-reactivity with recombinant human IFITM3 is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.