Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse IFN-alpha 2/IFNA2 Antibody, R&D Systems™

Rat Monoclonal Antibody
$177.10 - $402.50
Specifications
Antigen | IFN-alpha 2 |
---|---|
Clone | 996548 |
Dilution | ELISA |
Applications | ELISA |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB101491SP
![]() |
R&D Systems
MAB101491SP |
25 μg |
Each for $177.10
|
|
|||||
RD10000026
![]() |
R&D Systems
MAB101491100 |
100 μg |
Each for $402.50
|
|
|||||
Description
IFN-alpha 2 Monoclonal antibody specifically detects IFN-alpha 2 in Mouse samples. It is validated for ELISASpecifications
IFN-alpha 2 | |
ELISA | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
alpha-2a interferon, Ifa2, IFNA, IFNA2, IFNA2a, IFNA2b, IFNA2c, IFNAA, IFNalpha 2, IFN-alpha-2, IFN-alphaA, INFA2, interferon alpha 2b, interferon alpha A, interferon alpha-2, Interferon alpha-A, interferon, alpha 2, LeIF A, MGC125764, MGC125765 | |
Human embryonic kidney cell, HEK293-derived mouse IFN-alpha 2/IFNA2, Cys24-Glu190, Accession # P01573 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. |
996548 | |
ELISA | |
Unconjugated | |
Rat | |
Mouse | |
P01573 | |
3440 | |
IgG1 | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title