Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse IFN-alpha 4 Antibody, R&D Systems™

Rat Monoclonal Antibody
$200.50 - $432.50
Specifications
Antigen | IFN-alpha 4 |
---|---|
Clone | 1020125 |
Dilution | Immunohistochemistry 5-25 ug/mL |
Applications | Immunohistochemistry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB10259SP
![]() |
R&D Systems
MAB10259SP |
25 μg |
Each for $200.50
|
|
|||||
MAB10259100
![]() |
R&D Systems
MAB10259100 |
100 μg |
Each for $432.50
|
|
|||||
Description
IFN-alpha 4 Monoclonal specifically detects IFN-alpha 4 in Mouse samples. It is validated for Immunohistochemistry.Specifications
IFN-alpha 4 | |
Immunohistochemistry 5-25 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
IFNA4, IFNalpha 4, IFN-alpha-4, IFN-alpha4a, INFA4, interferon alpha-4, Interferon alpha-4B, Interferon alpha-76, Interferon alpha-M1, interferon, alpha 4, MGC142200 | |
Human embryonic kidney cell HEK293-derived mouse IFN-alpha 4, Leu23-Glu186, Accession # P07351 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. |
1020125 | |
Immunohistochemistry | |
Unconjugated | |
Rat | |
Mouse | |
P07351 | |
3441 | |
IgG2a | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title