Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse IFN-alpha/beta R2 Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 2 publications
Supplier: R&D Systems AF1083SP
This item is not returnable.
View return policy
Description
IFN-alpha/beta R2 Polyclonal specifically detects IFN-alpha/beta R2 in Mouse samples. It is validated for Western Blot, Neutralization.Specifications
IFN-alpha/beta R2 | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
beta receptor10, human interferon alpha, human interferon alpha/beta receptor, IFNABR, IFN-alpha binding protein, IFN-alpha/beta receptor 2, IFN-alpha-REC, IFN-aR2, IFNAR2, IFNARB, IFNbR2, IFN-bR2, IFN-R, IFN-R-2, interferon (alpha, beta and omega) receptor 2, Interferon alpha binding protein, interferon alpha/beta receptor 2, interferon-alpha/beta receptor beta chain, Type I interferon receptor 2 | |
Goat | |
Affinity Purified | |
RUO | |
3455 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot, Neutralization | |
LYOPH | |
Western Blot 0.1 ug/mL, Neutralization 2-10 ug/mL | |
Q9D1R7 | |
IFNAR2 | |
Mouse myeloma cell line NS0-derived recombinant mouse IFN-alpha/beta R2 Ser22-Ala239 (Lys160Asn) Accession # Q9D1R7 | |
25 μg | |
Primary | |
Detects mouse IFN-alpha/beta R2 in direct ELISAs and Western blots. In Western blots, approximately 10% cross-reactivity with recombinant mouse (rm) IFN-gamma R2 and rmIL-20 R alpha is observed, 5% cross-reactivity with recombinant human (rh) IL-10 R beta , rmIFN-gamma RI, and rhIL-20 R beta is observed and less than 1% cross-reactivity with rhIFN-beta R alpha is observed. | |
Mouse | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction