Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse IFN-alpha/beta R2 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF1083
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF1083 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF1083 Supplier R&D Systems Supplier No. AF1083
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody has been used in 2 publications

IFN-alpha/beta R2 Polyclonal specifically detects IFN-alpha/beta R2 in Mouse samples. It is validated for Western Blot, Neutralization.

Specifications

Antigen IFN-alpha/beta R2
Applications Western Blot, Neutralization
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.1 ug/mL, Neutralization 2-10 ug/mL
Gene Accession No. Q9D1R7
Gene Alias beta receptor10, human interferon alpha, human interferon alpha/beta receptor, IFNABR, IFN-alpha binding protein, IFN-alpha/beta receptor 2, IFN-alpha-REC, IFN-aR2, IFNAR2, IFNARB, IFNbR2, IFN-bR2, IFN-R, IFN-R-2, interferon (alpha, beta and omega) receptor 2, Interferon alpha binding protein, interferon alpha/beta receptor 2, interferon-alpha/beta receptor beta chain, Type I interferon receptor 2
Gene Symbols IFNAR2
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse IFN-alpha/beta R2 Ser22-Ala239 (Lys160Asn) Accession # Q9D1R7
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3455
Test Specificity Detects mouse IFN-alpha/beta R2 in direct ELISAs and Western blots. In Western blots, approximately 10% cross-reactivity with recombinant mouse (rm) IFN-gamma R2 and rmIL-20 R alpha is observed, 5% cross-reactivity with recombinant human (rh) IL-10 R beta , rmIFN-gamma RI, and rhIL-20 R beta is observed and less than 1% cross-reactivity with rhIFN-beta R alpha is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.