Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse IFN-beta Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. 324001
Change view
Click to view available options
Quantity:
20,000 U
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
324001 20,000 U
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 324001 Supplier R&D Systems Supplier No. 324001
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody has been used in 3 publications

IFN-beta Polyclonal specifically detects IFN-beta in Mouse samples. It is validated for Neutralization.

Specifications

Antigen IFN-beta
Applications Neutralization
Classification Polyclonal
Conjugate Unconjugated
Dilution Neutralization
Gene Alias Fibroblast interferon, IFB, IFBIFNB, IFF, IFNB, IFNB1, IFNbeta, IFN-beta, interferon beta, interferon, beta 1, fibroblast, MGC96956
Gene Symbols IFNB1
Host Species Rabbit
Immunogen E. coli-derived recombinant mouse IFN-beta
Purification Method Antigen Affinity-purified
Quantity 20,000 U
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3456
Test Specificity Mouse IFN-beta
Target Species Mouse
Content And Storage After receipt, this product should be kept at -70°C or below for retention of full activity.Thaw product vial by incubation in cold tap water until just thawed û the contents of the tube should be apportioned in separate tubes so that freezing and thawing is kept to a minimum. Refreezing should be done on dry ice or in a dry ice/alcohol bath.
Form Serum
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.