Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Description
Includes
Capture antibody, detection antibody, recombinant standard, streptavidin conjugated to horseradish-peroxidase (Streptavidin-HRP) and development protocols
The Mouse IFN-beta DuoSet ELISA is a Solid Phase Sandwich ELISA that quantifies mouse IFN-beta.
Specifications
Specifications
| Product Type | ELISA Kit |
| Species | Mouse |
| Binding Target | IFN-beta |
| Format | 96-well Plates |
| For Use With (Application) | ELISA |
| Gene ID (Entrez) | 3456 |
| Sample Type | Cell Culture Supernatant, Serum, Plasma |
| Synonym | Fibroblast interferon, IFB, IFBIFNB, IFF, IFNB, IFNB1, IFNbeta, IFN-beta, interferon beta, interferon, beta 1, fibroblast, MGC96956 |
| Content And Storage | Store the unopened product at 2°C to 8°C. Do not use past expiration date. |
For Research Use Only.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?
