Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse IFN-gamma R1/CD119 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB10262
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB10262 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB10262 Supplier R&D Systems Supplier No. MAB10262
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

IFN-gamma R1/CD119 Monoclonal specifically detects IFN-gamma R1/CD119 in Mouse samples. It is validated for Neutralization, ELISA Capture (Matched Antibody Pair).

Specifications

Antigen IFN-gamma R1/CD119
Applications Neutralization, ELISA
Classification Monoclonal
Clone 170911
Concentration LYOPH
Conjugate Unconjugated
Dilution Neutralization 4-20 ng/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. NP_034641
Gene Alias AVP, type 2, CD119, CD119 antigen, FLJ45734, IFN-gamma receptor 1, IFNgammaR1, IFN-gamma-R1, IFNGR, IFN-gR1, IFNGR1, immune interferon receptor 1, interferon gamma receptor 1, interferon-gamma receptor alpha chain, type 2
Gene Symbols IFNGR1
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse IFN-gamma R1/CD119 Gly23-Asp253 (predicted) Accession # NP_034641
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3459
Test Specificity Detects mouse IFN-gamma R1/CD119 in ELISAs. In sandwich immunoassays, no cross-reactivity or interference with recombinant mouse (rm) IFN-alpha/beta R1, rmIFN-gamma R2, recombinant human (rh) IFN-gamma R1, rhIL-20 R alpha , rhIL-20 R beta , rmIL-20 R alpha , rhIL-22BP or rmIL-22BP is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.