Learn More
Mouse IFN-gamma R2 Alexa Fluor 647-conjugated Antibody, R&D Systems™

Description
Specifications
Specifications
| Antigen | IFN-gamma R2 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | MOB47 |
| Concentration | 0.2 mg/mL |
| Conjugate | Alexa Fluor 647 |
| Dilution | Flow Cytometry 0.25-1 ug/10^6 cells |
| Formulation | Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide |
| Gene Alias | AF-1, AF-1interferon-gamma receptor beta chain precursor10IFNGT1Interferon gamma receptor accessory factor 1, IFGR2, IFN-gamma receptor 2, IFNgammaR2, IFN-gamma-R2, IFN-gR2, IFNGR2, IFNGT1, interferon gamma receptor 2, interferon gamma receptor 2 (interferon gamma transducer 1), interferon gamma receptor accessory factor-1, interferon gamma receptor beta chain, Interferon gamma transducer 1 |
| Gene Symbols | IFNGR2 |
| Show More |
For Research Use Only (RUO)
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.