Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse IFN-gamma R2 Alexa Fluor 750-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB773S100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB773S100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB773S100 Supplier R&D Systems Supplier No. FAB773S100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Hamster Monoclonal Antibody

IFN-gamma R2 Monoclonal specifically detects IFN-gamma R2 in Mouse samples. It is validated for Flow Cytometry.

Specifications

Antigen IFN-gamma R2
Applications Flow Cytometry
Classification Monoclonal
Clone MOB47
Concentration 0.2 mg/mL
Conjugate Alexa Fluor 750
Dilution Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
Gene Alias AF-1, AF-1interferon-gamma receptor beta chain precursor10IFNGT1Interferon gamma receptor accessory factor 1, IFGR2, IFN-gamma receptor 2, IFNgammaR2, IFN-gamma-R2, IFN-gR2, IFNGR2, IFNGT1, interferon gamma receptor 2, interferon gamma receptor 2 (interferon gamma transducer 1), interferon gamma receptor accessory factor-1, interferon gamma receptor beta chain, Interferon gamma transducer 1
Gene Symbols IFNGR2
Host Species Hamster
Immunogen Recombinant mouse IFN-gamma R2 Extracellular domain
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3460
Test Specificity Detects mouse IFN-gamma R2 extracellular domain in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant human (rh) IFN-gamma R2, recombinant mouse IFN-gamma R1, rhIFN-gamma R1, or rhIL-10 R beta is observed.
Target Species Mouse
Content And Storage Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Isotype IgG
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.