Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse IFN-gamma R2 Antibody, R&D Systems™

Hamster Monoclonal Antibody
Supplier: R&D Systems MAB773
This item is not returnable.
View return policy
Description
IFN-gamma R2 Monoclonal specifically detects IFN-gamma R2 in Mouse samples. It is validated for Western Blot, Flow Cytometry, Neutralization, CyTOF-ready.Specifications
IFN-gamma R2 | |
Monoclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
IFNGR2 | |
Recombinant mouse IFN-gamma R2 Extracellular domain | |
500 μg | |
Primary | |
Detects mouse IFN-gamma R2 extracellular domain in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant human (rh) IFN-gamma R2, recombinant mouse IFN-gamma R1, rhIFN-gamma R1, or rhIL-10 R beta is observed. | |
Mouse | |
Purified |
Western Blot, Flow Cytometry, Neutralization, CyTOF | |
MOB47 | |
Western Blot 1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, Neutralization 0.2-1 ug/mL, CyTOF-ready | |
AF-1, AF-1interferon-gamma receptor beta chain precursor10IFNGT1Interferon gamma receptor accessory factor 1, IFGR2, IFN-gamma receptor 2, IFNgammaR2, IFN-gamma-R2, IFN-gR2, IFNGR2, IFNGT1, interferon gamma receptor 2, interferon gamma receptor 2 (interferon gamma transducer 1), interferon gamma receptor accessory factor-1, interferon gamma receptor beta chain, Interferon gamma transducer 1 | |
Hamster | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
3460 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction