Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Mouse IgA ELISA Kit (Colorimetric)
SDP

Catalog No. NB112497 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB112497 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB112497 Supplier Novus Biologicals™ Supplier No. NBP300471

Mouse IgA ELISA Kit (Colorimetric) measures IgA in Serum, plasma, cell supernatant, and other biological fluids.

Specifications

Assay Range 15.63 to 1000 ng/mL
Assay Sensitivity 9.38 ng/mL
Product Type ELISA Kit (Colorimetric)
Sample Type Cell Culture Supernatant, Other Biological Fluids, Plasma, Serum
Specificity This kit recognizes Mouse IgA in samples. No significant cross-reactivity or interference between Mouse IgA and analogues was observed.
Target Species Mouse
For Use With (Application) ELISA
Gene ID (Entrez) 3493
Gene Symbol IGHA1
Quantity 1 Kit
Research Discipline Adaptive Immunity, Immunology
Storage Requirements Storage of components varies. See protocol for specific instructions.
Synonym Immunoglobulin A
Target IgA
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.