Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse IgD Antibody, R&D Systems™

Rat Monoclonal Antibody
Supplier: R&D Systems MAB9858SP
This item is not returnable.
View return policy
Description
IgD Monoclonal specifically detects IgD in Mouse samples. It is validated for Flow Cytometry, CyTOF-ready.Specifications
IgD | |
Monoclonal | |
Unconjugated | |
Lyophilized from a 0.2μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2μm filtered solution in PBS. | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
Primary | |
Reacts with the delta chain of Mouse IgD of all haplotypes and does not cross-react with other immunoglobulin isotypes. | |
Mouse | |
Supernatant |
CyTOF, Flow Cytometry | |
11-26C | |
Flow Cytometry 0.25μg/106 cells, CyTOF-ready | |
IgD, Igh-5, Ighd, Immunoglobulin D, Immunoglobulin Delta | |
Mouse IgD | |
25 μg | |
3495 | |
Reconstitute at 0.5mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction