Learn More
Mouse IgE Antibody (LO-ME-3), DyLight 405, Novus Biologicals™
SDP
Rat Monoclonal Antibody
Supplier:  Novus Biologicals NBP142334V
Catalog No. NBP142334V
Only null left
Add to Cart
Description
Description
Mouse IgE Monoclonal specifically detects Mouse IgE in Mouse samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Frozen.
Specifications
Specifications
Mouse IgE
Monoclonal
DyLight 405
50mM Sodium Borate with 0.05% Sodium Azide
Rat
Protein A or G purified
RUO
Mouse epsilon heavy chain of immunoglobulin (determined by immunodot).
Store at 4C in the dark.
IgG1 κ
ELISA, Immunohistochemistry, Immunohistochemistry (Frozen)
LO-ME-3
ELISA, Immunohistochemistry, Immunohistochemistry-Frozen
Immunoglobulin E, ImmunoglobulinE
The antibody reacts with the Mouse IgE antigen.
0.2 mL
Primary
Mouse
Purified
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.