Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Mouse IgE ELISA Kit (Colorimetric)
SDP

Catalog No. NB170632 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB170632 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB170632 Supplier Novus Biologicals™ Supplier No. NBP318786
Add to Cart
Add to Cart
This item is not returnable. View return policy

Colorimetric ELISA Kit

Specifications

Assay Range 125-8000 ng/mL
Assay Sensitivity 75 ng/mL
Product Type ELISA Kit (Colorimetric)
Sample Type Biological Fluids, Plasma, Serum
Specificity This kit recognizes Mouse IgE in samples. No significant cross-reactivity or interference between Mouse IgE and analogues was observed.
For Use With (Application) ELISA, Sandwich ELISA
Gene ID (Entrez) 3497
Gene Symbol IGHE
Quantity 1 Kit
Research Discipline Adaptive Immunity, Immunology
Storage Requirements Storage of components varies. See protocol for specific instructions.
Synonym IgE, immunoglobulin heavy constant epsilon
Target Mouse
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.