Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse IGFALS/ALS Antibody, R&D Systems™

Rat Monoclonal Antibody
Supplier: R&D Systems MAB1436
This item is not returnable.
View return policy
Description
ALS Monoclonal specifically detects ALS in Mouse samples. It is validated for Western Blot.Specifications
ALS | |
Monoclonal | |
LYOPH | |
Western Blot 2 ug/mL | |
P70389 | |
Igfals | |
Mouse myeloma cell line NS0-derived recombinant mouse IGFALS/ALS Thr28-Cys603 Accession # P70389 | |
500 μg | |
Primary | |
Detects mouse IGFALS/ALS in direct ELISAs and Western blots. | |
Mouse | |
Purified |
Western Blot | |
199410 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
ACLSD, ALS, Igfals | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
3483 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction