Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse IGFBP-1 Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems AF1240
This item is not returnable.
View return policy
Description
IGFBP-1 Polyclonal specifically detects IGFBP-1 in Mouse samples. It is validated for Western Blot, Immunohistochemistry.Specifications
IGFBP-1 | |
Polyclonal | |
Unconjugated | |
P47876 | |
IGFBP1 | |
Mouse myeloma cell line NS0-derived recombinant mouse IGFBP-1 Ala26-Asn272 Accession # P47876 | |
100 μg | |
Primary | |
Detects mouse IGFBP-1 in direct ELISAs and Western blots. In direct ELISAs, less than 10% cross-reactivity with recombinant human IGFBP-1 and less than 1% cross-reactivity with recombinant mouse (rm) IGFBP-4, and rmIGFBP-5 is observed. | |
Mouse | |
IgG |
Western Blot, Immunohistochemistry | |
LYOPH | |
Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL | |
alpha-pregnancy-associated endometrial globulin, amniotic fluid binding protein, binding protein-25, binding protein-26, binding protein-28, growth hormone independent-binding protein, hIGFBP-1, IBP1, IBP-1, IGF-binding protein 1, IGFBP1, IGFBP-1, IGF-BP25, insulin-like growth factor binding protein 1, insulin-like growth factor-binding protein 1, Placental protein 12, PP12AFBP | |
Goat | |
Affinity Purified | |
RUO | |
3484 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction