Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse IGFBP-2 Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 3 publications
$200.50 - $538.00
Specifications
Antigen | IGFBP-2 |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 0.1 ug/mL, Neutralization 30 ug/mL |
Applications | Western Blot, Neutralization |
Classification | Polyclonal |
Description
IGFBP-2 Polyclonal specifically detects IGFBP-2 in Mouse samples. It is validated for Western Blot, Neutralization.Specifications
IGFBP-2 | |
Western Blot 0.1 ug/mL, Neutralization 30 ug/mL | |
Polyclonal | |
Goat | |
Mouse | |
BP2, IBP2, IBP-2, IGF-binding protein 2, IGFBP2, IGFBP-2, IGF-BP53, insulin-like growth factor binding protein 2 (36kD), insulin-like growth factor binding protein 2, 36kDa, insulin-like growth factor-binding protein 2 | |
IGFBP2 | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Western Blot, Neutralization | |
Unconjugated | |
RUO | |
CAA57270 | |
3485 | |
Mouse myeloma cell line NS0-derived recombinant mouse IGFBP-2 Glu35-Gln305 Accession # CAA57270 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects mouse IGFBP-2 in direct ELISAs and Western blots. In direct ELISAs, approximately 35% cross-reactivity with recombinant human (rh) IGFBP-2 is observed and less than 1% cross-reactivity with rhIGFBP-1, rhIGFBP-3, rhIGFBP-4, rhIGFBP-5, recombinant mouse (rm) IGFBP-3, rmIGFBP-4, rmIGFBP-5, and rmIGFBP-6 is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title