Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse IGFBP-3 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF775
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF775 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF775 Supplier R&D Systems Supplier No. AF775
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody has been used in 3 publications

IGFBP-3 Polyclonal specifically detects IGFBP-3 in Mouse samples. It is validated for Western Blot, Simple Western.

Specifications

Antigen IGFBP-3
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.2 ug/mL, Simple Western 10 ug/mL
Gene Accession No. P47878
Gene Alias acid stable subunit of the 140 K IGF complex, binding protein 29, binding protein 53, growth hormone-dependent binding protein, IBP-3, IBP3BP-53, IGF-binding protein 3, IGFBP3, IGFBP-3, insulin-like growth factor binding protein 3, insulin-like growth factor-binding protein 3
Gene Symbols IGFBP3
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse IGFBP-3 (R&D Systems, Catalog # 775-B3) Pro22-Gln291 with Arg250Gln, Gln259Arg, Ser260Gly, Arg271Pro substitutions Accession # P47878
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3486
Test Specificity Detects mouse IGFBP-3 in direct ELISAs and Western blots. In direct ELISAs, approximately 5% cross-reactivity with recombinant human (rh) IGFBP-3 is observed and less than 1% cross-reactivity with rhIGFBP-1, rhIGFBP-2, rhIGFBP-4, rhIGFBP-5, rhIGFBP-6, and recombinant mouse IGFBP-6 is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.