Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse IGFBP-5 Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 3 publications
Supplier: R&D Systems AF578
This item is not returnable.
View return policy
Description
IGFBP-5 Polyclonal specifically detects IGFBP-5 in Mouse samples. It is validated for Western Blot, Immunohistochemistry, ELISA Capture (Matched Antibody Pair).Specifications
IGFBP-5 | |
Polyclonal | |
Unconjugated | |
Q3UQV0 | |
IGFBP5 | |
Mouse myeloma cell line NS0-derived recombinant mouse IGFBP-5 Val15-Glu271 Accession # Q3UQV0 | |
100 μg | |
Primary | |
Detects mouse IGFBP-5 in ELISAs and Western blots. In sandwich ELISAs, less than 3% cross-reactivity with recombinant human IGFBP-5 is observed, and less than 0.3% cross-reactivity with recombinant mouse (rm) IGF-I, rmIGF-II, rmIGFBP-1, rmIGFBP-2, rmIGFBP-3, and rmIGFBP-6 is observed. | |
Mouse | |
IgG |
Western Blot, Immunohistochemistry, ELISA | |
LYOPH | |
Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL, ELISA Capture (Matched Antibody Pair) 0.2-0.8 ug/mL | |
IBP5IBP-5, IGF-binding protein 5, IGFBP5, IGFBP-5, insulin-like growth factor binding protein 5, insulin-like growth factor-binding protein 5 | |
Goat | |
Affinity Purified | |
RUO | |
3488 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction