Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse IGFBP-6 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB776
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB776 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB776 Supplier R&D Systems Supplier No. MAB776
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

IGFBP-6 Monoclonal specifically detects IGFBP-6 in Mouse samples. It is validated for Western Blot.

Specifications

Antigen IGFBP-6
Applications Western Blot
Classification Monoclonal
Clone 122316
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P47880
Gene Alias IBP6, IBP-6, IGF binding protein 6, IGF-binding protein 6, IGFBP6, IGFBP-6, insulin-like growth factor binding protein 6, insulin-like growth factor-binding protein 6
Gene Symbols IGFBP6
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse IGFBP-6 Gly24-Gly238 (predicted) Accession # P47880
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3489
Test Specificity Detects mouse IGFBP-6 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human (rh) IGFBP-1, rhIGFBP-2, recombinant mouse (rm) IGFBP-2, rhIGFBP-3, rmIGFBP-3, rhIGFBP-4, rhIGFBP-5, rmIGFBP-5, and rhIGFBP-6 is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.