Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse IGFBP-rp1/IGFBP-7 Antibody, R&D Systems™

Rat Monoclonal Antibody has been used in 2 publications
$200.50 - $502.50
Specifications
Antigen | IGFBP-rp1/IGFBP-7 |
---|---|
Clone | 292917 |
Concentration | LYOPH |
Dilution | Immunohistochemistry 8-25 ug/mL |
Applications | Immunohistochemistry |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB21201SP
![]() |
R&D Systems
MAB21201SP |
25 μg |
Each for $200.50
|
|
|||||
MAB21201
![]() |
R&D Systems
MAB21201 |
100 μg |
Each for $502.50
|
|
|||||
Description
IGFBP-rp1/IGFBP-7 Monoclonal specifically detects IGFBP-rp1/IGFBP-7 in Mouse samples. It is validated for Immunohistochemistry.Specifications
IGFBP-rp1/IGFBP-7 | |
LYOPH | |
Immunohistochemistry | |
Unconjugated | |
Rat | |
Mouse | |
A0A5E1 | |
3490 | |
S. frugiperda insect ovarian cell line Sf 21-derived recombinant mouse IGFBP-rp1/IGFBP-7 Ser28-Leu281 (Arg94Lys) Accession # A0A5E1 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects mouse IGFBP-rp1/IGFBP-7 in direct ELISAs. |
292917 | |
Immunohistochemistry 8-25 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
angiomodulin, FSTL2, IBP-7, IGFBP7, IGFBP-7, IGFBP-7PGI2-stimulating factor, IGFBP-7v, IGFBPrp1, IGFBP-rP1, insulin-like growth factor binding protein 7, insulin-like growth factor-binding protein 7, Mac25, MAC25 protein, MAC25IGF-binding protein 7, Prostacyclin-stimulating factor, PSF, PSFAGM, TAF, Tumor-derived adhesion factor | |
IGFBP7 | |
IgG1 | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title