Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Mouse IgG ELISA Kit (Colorimetric)
SDP

Catalog No. NB600621KT Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB600621KT 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB600621KT Supplier Novus Biologicals™ Supplier No. NBP2600621KIT
Add to Cart
Add to Cart

Colorimetric ELISA Kit is useful in study of Cell Adhesion

The Mouse IgG ELISA Kit (Colorimetric) measures mouse IgG in serum, plasma, cell culture supernates, urine, tissue homogenates.

Specifications

Assay Range 125 to 8000ng/mL
Assay Sensitivity 29ng/mL
Product Type ELISA Kit (Colorimetric)
Sample Type Cell Culture Supernatant, Plasma, Serum, Tissue Homogenates, Urine
Specificity Detection of mouse IgG
Target Species Mouse
For Use With (Application) ELISA
Kit Contents Assay plate, Standard, HRP-conjugate, Sample Diluents, Wash Buffer (25X concentrate), TMB Substrate, Stop Solution, Adhesive Strip (For 96 wells), Instruction manual
Quantity 1 Kit
Regulatory Status RUO
Research Discipline Immunology
Sample Volume 50 to 100 μL
Storage Requirements Store the unopened product at 2° to 8°C. Protect from light. Do not use past expiration date.
Synonym Immunoglobulin heavy constant gamma 1 (G1m marker)
Target IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.