Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse IgG Goat anti-Mouse, Clone: RMG07 (Biotin), Abnova™

Catalog No. 89035299 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-035-299 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-035-299 Supplier Abnova Corporation Supplier No. MAB14947
Add to Cart
Add to Cart

Goat Monoclonal Antibody

Goat monoclonal antibody raised against native mouse IgG.

  • High specificity
  • High reactivity

Specifications

Antigen Mouse IgG
Applications ELISA, Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Immunoprecipitation
Classification Monoclonal
Clone RMG07
Conjugate Biotin
Description Goat monoclonal antibody raised against native mouse IgG.
Dilution ELISA (0.05-1 ug/mL) Flow Cytometry Immunocytochemistry Immunohistochemistry (0.5-2 ug/mL) Immunoprecipitation The optimal working dilution should be determined by the end user.
Formulation In PBS (50% glycerol, 1% BSA, 0.09% sodium azide)
Gene Accession No. P01868
Gene Alias IgG1/Igh-4/VH7183
Gene Symbols Ighg1
Host Species Goat
Immunogen Native purified mouse IgG.
Purification Method Protein G purification
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16017
Target Species Mouse
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.