Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse IgG1 Isotype Control (MG1), Janelia Fluor 549, Novus Biologicals™
SDP

Catalog No. NB001317 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB001317 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB001317 Supplier Novus Biologicals Supplier No. NBP197005JF549
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

IgG1 Monoclonal specifically detects IgG1 in Mouse samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence, Fluorophore-linked immunosorbent assay.

Specifications

Antigen IgG1
Applications Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone MG1
Conjugate Janelia Fluor 549
Dilution Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence, Fluorophore-linked immunosorbent assay
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias IgG1, Igh-4, RGD1359539, VH7183
Host Species Mouse
Purification Method Selective Precipitation
Quantity 0.5 mL
Primary or Secondary Isotype Control (Primary)
Test Specificity Isotyping assay resulted the material is non-reactive with antisera to mouse IgG2a, IgG2b, IgG3, IgM, and IgA. Fluorescein Conjugated Mouse IgG1 was proven by Dot Blot.
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.