Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Mouse IgG1-k Negative Control, Azide Free, Mouse, Unlabeled, Clone: MOPC-21,

Catalog No. MABF1081ZMI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
MABF1081ZMI 100 μg Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABF1081ZMI Supplier MilliporeSigma™ Supplier No. MABF1081Z
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The MOPC-21 immunoglobulin is useful as an isotype-matched control. The MOPC-21 immunolglobulin has an unknown binding specificity and is used as an isotype control for mouse IgG1 antibodies.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Mouse IgG1k
Applications Flow Cytometry
Classification Monoclonal
Clone MOPC-21
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Purified Mouse monoclonal containing 10mM NaH2PO4, 150mM NaCl, pH7.2. This monoclonal antibody was purified from tissue culture supernatant via affinity chromatography.
Gene IgG1-k
Host Species Mouse
Immunogen Immunogen information for this antibody is proprietary information.
Purification Method Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Research Discipline Inflammation & Immunology
Primary or Secondary Primary
Content And Storage Stable for 1 year at 2°-8°C from date of receipt. Note: It is recommended to store the product undiluted at 2°-8°C and protected from prolonged exposure to light. Do not freeze.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.