Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse IgG1 Kappa Light Chain, Clone: P3.6.2.8.1, FITC, Isotype control, Novus Biologicals™
SDP

Catalog No. NBP14395601 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.05 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP14395601 0.1 mg
NBP14395605 0.05 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP14395601 Supplier Novus Biologicals Supplier No. NBP1439560.1MG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

IgG1 Kappa Monoclonal specifically detects IgG1 Kappa in Mouse samples. It is validated for Flow Cytometry.

Specifications

Antigen IgG1 Kappa Light Chain
Applications Flow Cytometry
Classification Monoclonal
Clone P3.6.2.8.1
Concentration 0.5 mg/mL
Conjugate FITC
Dilution Flow Cytometry 1:100 - 1:2000
Formulation Aqueous buffer, may contain carrier protein/stabilizer (proprietary). with 0.09% Sodium Azide
Host Species Mouse
Quantity 0.1 mg
Primary or Secondary Primary
Test Specificity This monoclonal mouse IgG1, kappa immunoglobulin is useful as an isotype control.
Target Species Mouse
Content And Storage Store at 4C in the dark.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.