Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse IgG1 Kappa Light Chain, Clone: P3.6.2.8.1, Isotype control, Novus Biologicals™
SDP

Catalog No. NBP1433195 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.05 mg
0.5 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP1433195 0.5 mg
NBP1433195M 0.05 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP1433195 Supplier Novus Biologicals Supplier No. NBP1433190.5MG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody has been used in 2 publications

IgG1 Kappa Light Chain Monoclonal specifically detects IgG1 Kappa Light Chain in Mouse samples. It is validated for Control, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blot.

Specifications

Antigen IgG1 Kappa Light Chain
Applications Control, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone P3.6.2.8.1
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:100 - 1:2000, Flow Cytometry 1:10 - 1:1000, Immunoprecipitation 1:10 - 1:500
Formulation Aqueous buffer, may contain carrier protein/stabilizer (proprietary). with 0.09% Sodium Azide
Host Species Mouse
Purification Method Affinity Purified
Quantity 0.5 mg
Primary or Secondary Primary
Test Specificity This monoclonal mouse IgG1, kappa immunoglobulin is useful as an isotype control.
Target Species Mouse
Content And Storage Store at 4C in the dark.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.