Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse IgG1 PE-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB9627P100
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB9627P-100 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB9627P-100 Supplier R&D Systems Supplier No. FAB9627P100
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Monoclonal Antibody

IgG1 Monoclonal specifically detects IgG1 in Mouse samples. It is validated for Flow Cytometry.

Specifications

Antigen IgG1
Applications Flow Cytometry
Classification Monoclonal
Clone 2213C
Conjugate PE
Dilution Flow Cytometry 10μL/106 cells
Formulation Supplied in a saline solution containing BSA and Sodium Azide.
Gene Alias IgG1, Igh-4, RGD1359539, VH7183
Host Species Rabbit
Immunogen Mouse IgG
Purification Method Protein A or G purified from cell culture supernatant
Quantity 100 Tests
Primary or Secondary Primary
Test Specificity Detects mouse IgG1 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with mouse IgG2B, mouse IgG2A, and mouse IgG3 was observed.
Target Species Mouse
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Form Supernatant
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.